Skip to Content
Merck
All Photos(1)

Key Documents

SML2834

Sigma-Aldrich

SGC-CAMKK2-1

≥98% (HPLC)

Synonym(s):

2-Cyclopentyl-4-(5-(m-tolyl)furo[2,3-b]pyridin-3-yl)benzoic acid, YL-36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23NO3
CAS Number:
Molecular Weight:
397.47
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC(C2=CN=C(OC=C3C4=CC(C5CCCC5)=C(C(O)=O)C=C4)C3=C2)=CC=C1

InChI

1S/C26H23NO3/c1-16-5-4-8-18(11-16)20-13-23-24(15-30-25(23)27-14-20)19-9-10-21(26(28)29)22(12-19)17-6-2-3-7-17/h4-5,8-15,17H,2-3,6-7H2,1H3,(H,28,29)

InChI key

TXIYVFVMXNFNRX-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective CaM kinase kinase (CaMKK) inhibitor with selective affinity toward CAMKK1/2 over other kinases.
SGC-CAMKK2-1 (YL-36) is a potent and selective CaM kinase kinase (CaMKK) inhibitor (CAMKK2 IC50 = 30 nM) that inhibits C4-2 cellular AMPK phosphorylation (IC50 = 1.6 μM) with selective affinity toward CAMKK1/2 over other kinases (404-kinase panel kinome-wide Discoverx screen).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service