SML2726
Endosidin9
≥98% (HPLC)
Synonym(s):
5-Bromo-N-(4-Nitrophenyl)-2-thiophenesulfonamide, ES9
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(Nc2ccc(cc2)[N+](=O)[O-])c1[s]c(cc1)Br
InChI
1S/C10H7BrN2O4S2/c11-9-5-6-10(18-9)19(16,17)12-7-1-3-8(4-2-7)13(14)15/h1-6,12H
Biochem/physiol Actions
Endosidin9 (ES9) is a potent inhibitor of clathrin-mediated endocytosis (CME) in Arabidopsis thaliana, HeLa cells and Drosophila neurons. It binds directly to N-terminal domain of clathrin heavy chain. Also, Endosidin9 is a mitochondrial uncoupler that displays protonophore activities leading to cytoplasm acidification.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature chemical biology, 15(6), 641-649 (2019-04-24)
Clathrin-mediated endocytosis (CME) is a highly conserved and essential cellular process in eukaryotic cells, but its dynamic and vital nature makes it challenging to study using classical genetics tools. In contrast, although small molecules can acutely and reversibly perturb CME
Nature communications, 7, 11710-11710 (2016-06-09)
ATP production requires the establishment of an electrochemical proton gradient across the inner mitochondrial membrane. Mitochondrial uncouplers dissipate this proton gradient and disrupt numerous cellular processes, including vesicular trafficking, mainly through energy depletion. Here we show that Endosidin9 (ES9), a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service