Skip to Content
Merck
All Photos(1)

Key Documents

SML2723

Sigma-Aldrich

NVS-ZP7-4

≥98% (HPLC)

Synonym(s):

(S)-1-(2-((6-fluorobenzo[d]thiazol-2-yl)amino)-3-phenylpropyl)-1′H-spiro[piperidine-4,4′-quinazolin]-2′(3′H)-one, 1-[(2S)-2-[(6-fluoro-2-benzothiazolyl)amino]-3-phenylpropyl]-spiro[piperidine-4,4′(1′H)-quinazolin]-2′(3′H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28FN5OS
CAS Number:
Molecular Weight:
501.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1cc2[s]c(nc2cc1)N[C@H](CN4CCC5(NC(=O)Nc6c5cccc6)CC4)Cc3ccccc3

InChI

1S/C28H28FN5OS/c29-20-10-11-24-25(17-20)36-27(32-24)30-21(16-19-6-2-1-3-7-19)18-34-14-12-28(13-15-34)22-8-4-5-9-23(22)31-26(35)33-28/h1-11,17,21H,12-16,18H2,(H,30,32)(H2,31,33,35)/t21-/m0/s1

InChI key

FZOFDZMKSAUTHT-NRFANRHFSA-N

Biochem/physiol Actions

NVS-ZP7-4 is a potent inhibitor of the Notch signaling pathway that induces endoplasmic reticulum (ER) stress and induces apoptosis in T-ALL cell lines. NVS-ZP7-4 is a potent and selective inhibitor of SLC39A7 (ZIP7) zinc transporter that increases ER zinc levels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin Nolin et al.
Nature chemical biology, 15(2), 179-188 (2019-01-16)
The identification of activating mutations in NOTCH1 in 50% of T cell acute lymphoblastic leukemia has generated interest in elucidating how these mutations contribute to oncogenic transformation and in targeting the pathway. A phenotypic screen identified compounds that interfere with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service