Skip to Content
Merck
All Photos(1)

Key Documents

SML2527

Sigma-Aldrich

SW063058

≥98% (HPLC)

Synonym(s):

7-Bromo-5-(4-fluorophenyl)-1,3,4,5-tetrahydro-4-(2-iodobenzoyl)-2H-1,4-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15BrFIN2O2
CAS Number:
Molecular Weight:
565.17
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Ic1c(cccc1)C(=O)N2C(c4c(ccc(c4)Br)NC(=O)C2)c3ccc(cc3)F

InChI

1S/C22H15BrFIN2O2/c23-14-7-10-19-17(11-14)21(13-5-8-15(24)9-6-13)27(12-20(28)26-19)22(29)16-3-1-2-4-18(16)25/h1-11,21H,12H2,(H,26,28)

InChI key

CFXZDEOFOAXVPQ-UHFFFAOYSA-N

Biochem/physiol Actions

Autophagy inducer that selectively disrupts Beclin 1/Bcl-2 binding
SW063058 is an autophagy inducer that selectively disrupts Beclin 1/Bcl-2 binding, versus Bax/Bcl-2 binding. SW063058 acts by disrupting the binding of Bcl-2, a negative regulator of autophagy, to the BH3 domain of Beclin 1, which is involved in autophagy initiation. It does not inhibit Bcl-2 binding to the BH3 domain of the pro-apoptotic Bcl-2 family members BAX and BIM. In vitro experiments showed that SW063058 was not cytotoxic and induced autophagy without inducing apoptosis or other forms of cell death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service