Skip to Content
Merck
All Photos(1)

Key Documents

SML1909

Sigma-Aldrich

RS 23597-190

≥98% (HPLC)

Synonym(s):

3-(Piperidin-1-yl)propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, RS 23597, RS-23,597-190, RS-23597, RS-23597-190, RS23597, RS23597-190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23ClN2O3 · HCl
CAS Number:
Molecular Weight:
363.28
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[Cl-].Clc1c(cc(c(c1)C(=O)OCCCN2CCCCC2)OC)N.[H+]

InChI

1S/C16H23ClN2O3.ClH/c1-21-15-11-14(18)13(17)10-12(15)16(20)22-9-5-8-19-6-3-2-4-7-19;/h10-11H,2-9,18H2,1H3;1H

InChI key

QLZLBYYNMGQIAR-UHFFFAOYSA-N

Biochem/physiol Actions

RS 23597-190 is a high affinity 5-HT4-selective antagonist with in vitro and in vivo efficacy. RS 23597-190 is a 4-amino-5-chloro-2-methoxybenzoate that antagonizes 5-HT-mediated relaxation of carbachol-contracted rat oesphageal muscularis mucosae ex vivo (pA2 = 7.5) and prevents 5-HT-induced tachycardia (tachyarrhythmia) of anaesthetized micropigs in vivo (6 mg/kg RS 23597-190 and 3-10 μ/kg 5-HT via i.v.) by targeting 5-HT4 with high affinity (pKB = 7.8) and selectivity, while exhibiting no efficacy against 5-methoxytryptamine-mediated 5-HT3 activation in guinea-pig ileum and displaying much reduced or little affinity toward 5-HT3 (pKi = 5.7/mouse NG108-15 cells and pKB <5/guinea-pig ileum), dopamine receptors (pKi <4/D1 & <3/D2), muscarinic receptors (pKi <4.5 toward M1/2/3/4), 5-HT1A & 5-HT2 (pKi <5.2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingyi Wang et al.
Scientific reports, 6, 37336-37336 (2016-11-23)
5-HT has been reported to possess significant effects on cardiac activities, but activation of 5-HTR on the cell membrane failed to illustrate the controversial cardiac reaction. Because 5-HT constantly comes across the cell membrane via 5-HT transporter (5-HTT) into the
R M Eglen et al.
British journal of pharmacology, 110(1), 119-126 (1993-09-01)
1. The pharmacological properties of RS 23597-190 (3-(piperdine-1-yl)-propyl-4-amino-5-chloro-2-methoxy benzoate hydrochloride) have been studied in vitro and in vivo. 2. RS 23597-190 competitively antagonized 5-HT4 receptor-mediated relaxations of rat, carbachol precontracted oesophageal muscularis mucosae, (pA2 = 7.8 +/- 0.1; Schild slope
Mohammad Nasehi et al.
Behavioural brain research, 317, 272-278 (2016-10-28)
Functional interaction between cannabinoid and serotonin neuronal systems have been reported in different tasks related to memory assessment. The present study investigated the effect of serotonin 5-HT4 agents into the dorsal hippocampus (the CA1 region) on spatial and object novelty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service