SML1699
(R)-ZINC-3573
≥98% (HPLC)
Synonym(s):
(R)-ZINC72453573, (R)-N,N-Dimethyl-1-(5-phenylpyrazolo[1,5-a]pyrimidin-7-yl)-3-pyrrolidinamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H21N5
CAS Number:
Molecular Weight:
307.39
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 5 mg/mL, clear (warmed)
storage temp.
2-8°C
SMILES string
CN([C@H](CC1)CN1C2=CC(C3=CC=CC=C3)=NC4=CC=NN24)C
Biochem/physiol Actions
(R)-ZINC-3573 is a selective agonist probe of the orphan receptor Mas-related G-protein coupled receptor member X2 (MRGPRX2), a G-protein coupled receptor (GPCR) selectively expressed in mast cells and dorsal root and trigeminal ganglia primary sensory neurons of primates. Lack of selective and potent probes has made it difficult to determine MRGPRX2 funtion. (R)-ZINC-3573 was shown to be a potent and selective agonist with an EC50 value of 760 nM for MRGPRX2 and little activity for MRGPRX1 or over 350 other GPCRs and 97 kinases tested. (R)-ZINC-3573 activates endogenous MRGPRX2 expressed in a human mast cell line and induced degranulation and calcium release. (S)-ZINC-3573 is inactive and can be used as a control.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
In silico design of novel probes for the atypical opioid receptor MRGPRX2
Lansu, K.
Nature Chemical Biology (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service