Skip to Content
Merck
All Photos(1)

Key Documents

SML0522

Sigma-Aldrich

JM6

≥97% (HPLC)

Synonym(s):

2-(3,4-Dimethoxybenzenesulfonylamino)-4-(3-nitrophenyl)-5-(piperidin-1-yl)methylthiazole, 3,4-Dimethoxy-N-[4-(3-nitrophenyl)-5-(1-piperidinylmethyl)-2-thiazolyl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N4O6S2
CAS Number:
Molecular Weight:
518.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)S(=O)(=O)Nc2nc(c(CN3CCCCC3)s2)-c4cccc(c4)[N+]([O-])=O

InChI

1S/C23H26N4O6S2/c1-32-19-10-9-18(14-20(19)33-2)35(30,31)25-23-24-22(16-7-6-8-17(13-16)27(28)29)21(34-23)15-26-11-4-3-5-12-26/h6-10,13-14H,3-5,11-12,15H2,1-2H3,(H,24,25)

InChI key

AOZLMMDVJOGGEQ-UHFFFAOYSA-N

Biochem/physiol Actions

JM6 is a prodrug inhibitor of kynurenine 3-monooxygenase (KMO) that increases kynurenic acid levels and reduces extracellular glutamate in the brain. JM6 prevents spatial memory deficits, anxiety-related behavior, and synaptic loss in mouse model of Alzheimer disease.
JM6 is a prodrug inhibitor of kynurenine 3-monooxygenase (KMO); Ro 61-8048 analog.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewelina Rojewska et al.
Neuropharmacology, 102, 80-91 (2015-11-03)
Recent studies have highlighted the involvement of the kynurenine pathway in the pathology of neurodegenerative diseases, but the role of this system in neuropathic pain requires further extensive research. Therefore, the aim of our study was to examine the role

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service