Skip to Content
Merck
All Photos(1)

Key Documents

SMB01054

Sigma-Aldrich

Umbelliprenin

≥90% (LC/MS-ELSD)

Synonym(s):

7-Farnesyloxycoumarin, umbelliprenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O3
CAS Number:
Molecular Weight:
366.49
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

366.49

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C24H30O3/c1-18(2)7-5-8-19(3)9-6-10-20(4)15-16-26-22-13-11-21-12-14-24(25)27-23(21)17-22/h7,9,11-15,17H,5-6,8,10,16H2,1-4H3/b19-9+,20-15+

InChI key

GNMUGVNEWCZUAA-WOWYBKFKSA-N

General description

Umbelliprenin is a sesquiterpene coumarin commonly found in plants such as Thamnosma montana, Scabiosa comosa, Ferula iliensis, and Palicourea alpina. Current research suggests that this naturally occurring metabolite may possess various biological activities, including anti-inflammatory, antioxidant, antitumor, and anticancer properties.

Application

Umbelliprenin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Umbelliprenin might be valuable as a cancer chemopreventive agent.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrdad Iranshahi et al.
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP), 18(5), 412-415 (2009-06-18)
Umbelliprenin is a prenylated compound, which belongs to the class of sesquiterpene coumarins. In continuation of our earlier in-vitro finding, we determined to assess the cancer chemopreventive activity of umbelliprenin in vivo by using a two-stage carcinogenesis assay of mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service