Skip to Content
Merck
All Photos(1)

Key Documents

SMB00511

Sigma-Aldrich

Skimmin

≥90% (LC/MS-UV)

Synonym(s):

7-Hydroxycoumarin-7-glucoside, Umbelliferone glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O8
CAS Number:
Molecular Weight:
324.28
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O)O)CO)OC2=CC(O3)=C(C=C2)C=CC3=O

InChI

1S/C15H16O8/c16-6-10-12(18)13(19)14(20)15(23-10)21-8-3-1-7-2-4-11(17)22-9(7)5-8/h1-5,10,12-16,18-20H,6H2/t10-,12-,13+,14-,15-/m1/s1

InChI key

VPAOSFFTKWUGAD-TVKJYDDYSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Renal protecting, anti-inflammation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sen Zhang et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 819296-819296 (2013-08-31)
Skimmin is one of the major pharmacologically active molecules present in Hydrangea paniculata, a medical herb used in the traditional Chinese medicine as an anti-inflammatory agent. In the current study, we attempted to investigate its renoprotective activity and underlying mechanisms
Sen Zhang et al.
European journal of pharmacology, 692(1-3), 78-83 (2012-06-06)
Skimmin, a major active ingredient from Hydrangea paniculata, was considered to have the possible preventive effect on the progression of diabetic nephropathy based on the traditional Chinese medicine. The present study aimed to assess this preventive activity in a rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service