Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504095

Sigma-Aldrich

Anti-phospho-p38 MAPK (pThr179+Tyr181) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr179/pTyr181)

Gene Information

human ... MAPK14(1432)

Immunogen

The antiserum was produced against synthesized peptide derived from human p38 MAPK around the phosphorylation site of Thr179 and Tyr181.

Immunogen Range: 151-200

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Cao et al.
Microbial pathogenesis, 131, 120-127 (2019-03-27)
Enterovirus 71 (EV71) is the main causative agent of hand, foot, and mouth disease (HFMD), which has high morbidity and mortality. It mainly threatens children under six years of age. Because of a poor understanding of its pathogenesis, there are
Yingxiao Fu et al.
Molecular medicine reports, 12(5), 6939-6945 (2015-09-04)
The present study aimed to determine whether the mitogen‑activated protein kinase (MAPK) signaling pathway is involved in the osteoprotegerin (OPG)‑mediated inhibition of osteoclast differentiation and maturation. RAW264.7 cells were incubated with macrophage colony‑stimulating factor (M‑CSF) and receptor activator of nuclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service