Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200841

Sigma-Aldrich

Anti-Syntaxin antibody, Mouse monoclonal

clone HPC-1, purified from hybridoma cell culture

Synonym(s):

Anti-STX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

purified from hybridoma cell culture

Quality Level

clone

HPC-1, monoclonal

form

liquid

species reactivity

monkey, rat, human, dog, mouse, bovine

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.25-0.5 μg/mL using human neuroblastoma SHSY5Y cell lysate.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Syntaxins are a family of membrane-integrated proteins that are instrumental in exocytosis of vesicles.6 The syntaxin family consists of 15 genes in mammals and 7 in yeast, and there are syntaxin-like sequences in all eukaryotes examined.7 All mammalian syntaxins, with the exception of syntaxin 11, are transmembrane proteins anchored by their C-terminal tails.7
Syntaxin-1(also known as HPC-1 antigen or Stx1),8-9 a 35 kDa molecule with carboxyl-terminal membrane anchor, is a synaptic protein identified by its ability to interact with the synaptic vesicle protein synaptotagmin

Specificity

Monoclonal Anti-Syntaxin antibody specifically recognizes membrane protein syntaxin, expressed predominantly in neuronal tissues.2,3 The antibody reacts with syntaxin from human, mouse and rat origin. The antibody has also been used to isolate syntaxin cDNA clones by screening a λgt 11 library from rat hippocampus.3

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (~35 kDa), Immunohistochemistry4 and Immunofluorescence5.

Biochem/physiol Actions

Syntaxin-1 protein is an essential component of the synaptic vesicle soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) complex and is essential for neurotransmission.10 It forms a four-helical bundle complex with proteins SNAP-25 and VAMP2 that drives fusion of vesicular and plasma membranes.6,11

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanilla Hua Shi et al.
Journal of neurochemistry, 156(5), 614-623 (2020-08-28)
The t-soluble NSF-attachment protein receptor protein Syntaxin-1a (Stx-1a) is abundantly expressed at pre-synaptic terminals where it plays a critical role in the exocytosis of neurotransmitter-containing synaptic vesicles. Stx-1a is phosphorylated by Casein kinase 2α (CK2α) at Ser14, which has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service