SAB4200623
Anti-C9orf72 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
3110043O21Rik, AI840585, ALSFTD, C9orf72, FLJ11109, FTDALS, RGD1359108, RP23-307M2.1, zgc:100846
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
form
buffered aqueous solution
mol wt
~55 kDa
species reactivity
human
concentration
~1 mg/mL
technique(s)
immunoblotting: 0.3-0.6 μg/mL using whole extracts of T98G cells
immunocytochemistry: 4-6 μg/mL using SH-SY-5Y cells
immunohistochemistry: 10 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human colon sections
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... C9orf72(203228)
General description
Specificity
Immunogen
Application
- immunoblotting
- immunocytochemistry
- immunohistochemistry
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service