Skip to Content
Merck
All Photos(1)

Key Documents

SAB2106887

Sigma-Aldrich

Anti-DGAT2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARAT, Anti-GS1999FULL, Anti-HMFN1045

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
bovine, dog, rabbit, horse, guinea pig, mouse, human, rat
citations:
1
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

bovine, dog, rabbit, horse, guinea pig, mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGAT2(84649)
mouse ... Dgat2(67800)

Immunogen

The immunogen for anti-DGAT2 antibody: synthetic peptide derected towards the C terminal of human DGAT2

Biochem/physiol Actions

Dgat2 is an essential acyltransferase that catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol and fatty acyl CoA as substrates. It is required for synthesis and storage of intracellular triglycerides. It probably plays a central role in cytosolic lipid accumulation.

Sequence

Synthetic peptide located within the following region: LMSGGICPVNRDTIDYLLSKNGSGNAIIIVVGGAAESLSSMPGKNAVTLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirinat Pengnet et al.
Frontiers in pharmacology, 13, 1049818-1049818 (2023-01-03)
Background: Excessive fructose consumption causes hepatic lipid accumulation via increased triglyceride (TG) synthesis, leading to the development and progression of non-alcoholic fatty liver disease (NALFD). Naringin, a flavanone glycoside found in citrus fruit, has antioxidant and hypolipidemic properties. Therefore, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service