Skip to Content
Merck
All Photos(2)

Key Documents

SAB1408259

Sigma-Aldrich

Anti-MRGPRX2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MRGX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37.1 kDa

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Mas-related G protein-coupled receptor X2 (MRGPRX2) belonging to the class A GPCR/Mas-related gene family is expressed in sensory neurons, mast cells, and keratinocytes. It has seven transmembrane (TM) α-helices. MRGPRX2 mRNA is found at a high level in the skin. It is also detected in adipose tissue, esophagus, urinary bladder, and lungs. MRGPRX2 gene with two-exon is mapped on human chromosome 11p15.

Immunogen

MRGPRX2 (NP_473371.1, 1 a.a. ~ 330 a.a) full-length human protein.

Sequence
MDPTTPAWGTESTTVNGNDQALLLLCGKETLIPVFLILFIALVGLVGNGFVLWLLGFRMRRNAFSVYVLSLAGADFLFLCFQIINCLVYLSNFFCSISINFPSFFTTVMTCAYLAGLSMLSTVSTERCLSVLWPIWYRCRRPRHLSAVVCVLLWALSLLLSILEGKFCGFLFSDGDSGWCQTFDFITAAWLIFLFMVLCGSSLALLVRILCGSRGLPLTRLYLTILLTVLVFLLCGLPFGIQWFLILWIWKDSDVLFCHIHPVSVVLSSLNSSANPIIYFFVGSFRKQWRLQQPILKLALQRALQDIAEVDHSEGCFRQGTPEMSRSSLV

Biochem/physiol Actions

Mas-related G protein-coupled receptor X2 (MRGPRX2) expressed on mast cells acts as an important receptor for pseudo-allergic responses induced by drugs. It is involved in the evolution of the human nervous system. It is also crucial to mast cells. MRGPRX2 gene mutation impairs the risk of anaphylactic actions facilitated by the MRGPRX2 receptor.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haihui Wu et al.
Progress in histochemistry and cytochemistry, 50(1-2), 11-17 (2015-06-25)
Mas-related genes (Mrgs) belong to a large family of G protein-coupled receptor genes found in rodents. Human MRGX proteins are G protein-coupled 7-transmembrane proteins sharing 41-52% amino acid identity with each other, but have no orthologs in rodents. MrgX2 is
Chalatip Chompunud Na Ayudhya et al.
International journal of molecular sciences, 22(10) (2021-06-03)
The neuropeptide substance P (SP) mediates neurogenic inflammation and pain and contributes to atopic dermatitis in mice through the activation of mast cells (MCs) via Mas-related G protein-coupled receptor (GPCR)-B2 (MrgprB2, human ortholog MRGPRX2). In addition to G proteins, certain
Grzegorz Porebski et al.
Frontiers in immunology, 9, 3027-3027 (2019-01-09)
The human ortholog MRGPRX2 and the mice ortholog, Mrgprb2 are activated by basic secretagogues and neurokinins. A number of commonly used small-molecule drugs (e.g., neuromuscular blocking agents, fluoroquinolones, vancomycin) have been recently shown to activate these receptors under in vitro
Di Wei et al.
Immunopharmacology and immunotoxicology, 43(1), 77-84 (2020-12-18)
Clozapine is one of the most widely used second-generation antipsychotics in clinic. However, allergy-like symptoms such as rash and angioedema have been reported frequently, and the mechanism is still not clear. Mas-related G protein-coupled receptor X2 (MRGPRX2) expressed on mast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service