Skip to Content
Merck
All Photos(3)

Key Documents

SAB1406807

Sigma-Aldrich

Anti-PDLIM7 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

LMP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~49.8 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM7(9260)

General description

The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants. (provided by RefSeq)

Immunogen

PDLIM7 (NP_005442.2, 1 a.a. ~ 457 a.a) full-length human protein.

Sequence
MDSFKVVLEGPAPWGFRLQGGKDFNVPLSISRLTPGGKAAQAGVAVGDWVLSIDGENAGSLTHIEAQNKIRACGERLSLGLSRAQPVQSKPQKASAPAADPPRYTFAPSVSLNKTARPFGAPPPADSAPQQNGQPLRPLVPDASKQRLMENTEDWRPRPGTGQSRSFRILAHLTGTEFMQDPDEEHLKKSSQVPRTEAPAPASSTPQEPWPGPTAPSPTSRPPWAVDPAFAERYAPDKTSTVLTRHSQPATPTPLQSRTSIVQAAAGGVPGGGSNNGKTPVCHQCHKVIRGRYLVALGHAYHPEEFVCSQCGKVLEEGGFFEEKGAIFCPPCYDVRYAPSCAKCKKKITGEIMHALKMTWHVHCFTCAACKTPIRNRAFYMEEGVPYCERDYEKMFGTKCHGCDFKIDAGDRFLEALGFSWHDTCFVCAICQINLEGKTFYSKKDRPLCKSHAFSHV

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Ren et al.
Molecular cancer, 16(1), 117-117 (2017-07-12)
The primary issue arising from prostate cancer (PCa) is its high prevalence to metastasize to bone, which severely affects the quality of life and survival time of PCa patients. miR-210-3p is a well-documented oncogenic miRNA implicated in various aspects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service