R2633
Cuvette rack
Holds 12 cuvettes
Synonym(s):
rack tube, racks tube, tube racks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Recommended Products
material
polypropylene
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
L × W × H
4 5/8 in. × 4 1/4 in. × 4 3/8 in.
Looking for similar products? Visit Product Comparison Guide
General description
Special design rack. Only the corners of the cells touch the rack, greatly reducing the risk of damage to the optical surfaces.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service