Skip to Content
Merck
All Photos(1)

Key Documents

PZ0207

Sigma-Aldrich

PF-04620110

≥98% (HPLC)

Synonym(s):

trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]-cyclohexaneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N4O4
CAS Number:
Molecular Weight:
396.44
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

room temp

SMILES string

Nc1ncnc2OCCN(C(=O)c12)c3ccc(cc3)[C@@H]4CC[C@H](CC4)CC(O)=O

InChI

1S/C21H24N4O4/c22-19-18-20(24-12-23-19)29-10-9-25(21(18)28)16-7-5-15(6-8-16)14-3-1-13(2-4-14)11-17(26)27/h5-8,12-14H,1-4,9-11H2,(H,26,27)(H2,22,23,24)/t13-,14-

InChI key

GEVVQZHMFVFGLN-HDJSIYSDSA-N

Biochem/physiol Actions

PF-04620110 is an orally active, selective and potent DGAT1 (Acyl-CoA:diacylglycerol acyltransferase 1) inhibitor that inhibits triacylglycerol synthesis in cells and in rodents.
PF-04620110 is known to regulate gut hormones. Inhibition of DGAT1 might serve as a good approach for the treatment of obesity and type 2 diabetes. DGAT1 inhibition might increase the oxidation of fatty acids, thus it is found to be protective against hepatic steatosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific role for acyl CoA: Diacylglycerol acyltransferase 1 (Dgat1) in hepatic steatosis due to exogenous fatty acids
Villanueva CJ, et al.
Hepatology, 50(2), 434-442 (2009)
Targeting Acyl-CoA: diacylglycerol acyltransferase 1 (DGAT1) with small molecule inhibitors for the treatment of metabolic diseases
Cao J, et al.
The Journal of biological chemistry, jbc-M111 (2011)
Targeting Acyl-CoA: diacylglycerol acyltransferase 1 (DGAT1) with small molecule inhibitors for the treatment of metabolic diseases
Cao J, et al.
The Journal of Biological Chemistry, jbc-M111 (2011)
New Therapeutic Strategies for Type 2 Diabetes: Small Molecule Approaches, 237-237 (2012)
Targeting Acyl-CoA: diacylglycerol acyltransferase 1 (DGAT1) with small molecule inhibitors for the treatment of metabolic diseases
Cao J, et al.
Test, jbc-M111 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service