Skip to Content
Merck
All Photos(1)

Key Documents

PZ0150

Sigma-Aldrich

SU5614

≥98% (HPLC)

Synonym(s):

(3Z)-5-Chloro-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylene]-1,3-dihydro-2H-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13ClN2O
CAS Number:
Molecular Weight:
272.73
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(\C=C2/C(=O)Nc3ccc(Cl)cc23)[nH]1

InChI

1S/C15H13ClN2O/c1-8-5-9(2)17-14(8)7-12-11-6-10(16)3-4-13(11)18-15(12)19/h3-7,17H,1-2H3,(H,18,19)/b12-7-

InChI key

XLBQNZICMYZIQT-GHXNOFRVSA-N

Biochem/physiol Actions

SU5614 is a FMS-like tyrosine kinase 3 (FLT3) inhibitor; selective inhibitor of VEGF and PDGF receptor tyrosine kinases.

Features and Benefits

This compound is featured on the PDGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoko Matsushima et al.
Scientific reports, 10(1), 188-188 (2020-01-15)
Anosmin-1 is a secreted glycoprotein encoded by the ANOS1 gene, and its loss of function causes Kallmann syndrome (KS), which is characterized by anosmia and hypogonadism due to olfactory bulb (OB) dysfunction. However, the physiological function of anosmin-1 remains to
Ayumi Yoshida et al.
Biology open, 4(9), 1063-1076 (2015-07-26)
Neuropilin-1 (NRP1) has been identified as a VEGF-A receptor. DJM-1, a human skin cancer cell line, expresses endogenous VEGF-A and NRP1. In the present study, the RNA interference of VEGF-A or NRP1 suppressed DJM-1 cell proliferation. Furthermore, the overexpression of
Manja Wobus et al.
Oncotarget, 6(36), 38804-38815 (2015-10-16)
Internal tandem duplications within the juxtamembrane region of the FMS-like tyrosine kinase receptor FLT3 (FLT3-ITD) represents one of the most common mutations in patients with acute myeloid leukemia (AML) which results in constitutive aberrant activation, increased proliferation of leukemic progenitors

Articles

We offers many products related to PDGFR for your research needs.

We offers many products related to PDGFR for your research needs.

We offers many products related to PDGFR for your research needs.

We offers many products related to PDGFR for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service