Skip to Content
Merck
All Photos(3)

Documents

PLA0127

Sigma-Aldrich

Rabbit anti-RNA Polymerase II Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

220kDa, DNA-directed RNA polymerase II largest subunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, POLR2, POLRA, RNA polymerase II 220 kd subunit, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1, polymerase (RNA) II (DNA directed) polypeptide A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:1,000- 1:10,000

accession no.

NP_000928.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

The epitope recognized by PLA0127 maps to the C-terminal repeat of human RNA Polymerase II (GeneID 5430).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

The RNA polymerase II subunit B1 (RPB1) is the largest subunit of the RNA polymerase II core complex. RNA polymerase II holoenzyme functions to transcribe DNA into messenger RNA in eukaryotes. RPB1 forms the DNA binding domain of the polymerase that forms a cleft that grabs the incoming DNA template to translocate the holoenzyme across the RNA-DNA hybrid.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanchang Liu et al.
Frontiers in bioengineering and biotechnology, 9, 735949-735949 (2021-12-07)
Bone marrow mesenchymal stem cells (BMSCs) play a critical role in bone formation and are extremely sensitive to external mechanical stimuli. Mechanical signals can regulate the biological behavior of cells on the surface of titanium-related prostheses and inducing osteogenic differentiation
Zhenhua Liu et al.
Journal of cellular and molecular medicine, 22(9), 4161-4170 (2018-07-06)
Oestrogen receptor ɑ (ERɑ) is overexpressed in two-thirds of all breast cancers and involves in development and breast cancer progression. Although ERɑ-positive breast cancer could be effective treated by endocrine therapy, the endocrine resistance is still an urgent clinical problem.
Y Xu et al.
Oncogene, 36(8), 1157-1166 (2016-08-16)
The heterogeneous breast cancers can be classified into different subtypes according to their histopathological characteristics and molecular signatures. Foxa1 expression is linked with luminal breast cancer (LBC) with good prognosis, whereas Twist1 expression is associated with basal-like breast cancer (BLBC)
Anirban Chakraborty et al.
The Journal of biological chemistry, 299(3), 102991-102991 (2023-02-10)
A growing body of evidence indicates that RNA plays a critical role in orchestrating DNA double-strand break repair (DSBR). Recently, we showed that homologous nascent RNA can be used as a template for error-free repair of double-strand breaks (DSBs) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service