Key Documents
P6679
Sigma-Aldrich
Microcapillary pipettes, positive displacement
volume range 1-5 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
glass
packaging
pack of 100 ea
manufacturer/tradename
Drummond Scientific 5-000-1001-X
capacity
1 - 5 μL
volume range
1-5 μL
General description
Calibrated glass microcapillary pipettes with separate stainless steel plunger wires. Not damaged by dry sterilization above 400 °F (205 °C). Pipette 50% glycerol solutions accurately without aerosol formation. Single-use disposable pipette and plunger to avoid cross-contamination.
Application
Designed specifically for use in PCR work
Packaging
Package contains 100 pipettes, 100 plunger wires, and one reusable ring-style handle.
Choose from one of the most recent versions:
Certificates of Analysis (COA)