I4028
Monoclonal Anti-Interleukin-18 Receptor antibody produced in mouse
clone 70625.111, purified immunoglobulin, lyophilized powder
Synonym(s):
Anti-IL-18 Receptor
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
70625.111, monoclonal
form
lyophilized powder
species reactivity
human
technique(s)
flow cytometry: suitable (2.5μg/SUP>106 cell)
immunocytochemistry: 8-25 μg/mL
immunohistochemistry: 8-25 μg/mL
isotype
IgG1
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... IL18R1(8809)
Related Categories
General description
Immunogen
Biochem/physiol Actions
Physical form
Preparation Note
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service