Skip to Content
Merck
All Photos(6)

Key Documents

HPA025826

Sigma-Aldrich

Anti-MRPL37 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-39S ribosomal protein L37, mitochondrial, Anti-L37mt, Anti-MRP-L37

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

SATWNRESLLLQVRGSGGARLSTKDPLPTIASREEIEATKNHVLETFYPISPIIDLHECNIYDVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL37(51253)

General description

The gene MRPL37 (mitochondrial ribosomal protein L37) is mapped to human chromosome 1p32.3. The gene encodes a mitochondrial ribosomal protein of the large subunit. The MRPL37 protein associates with MTO1 (mitochondrial translation optimization factor 1), a regulator of mitochondrial translation.

Immunogen

39S ribosomal protein L37, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76585

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedetta Ruzzenente et al.
Methods in molecular biology (Clifton, N.J.), 2224, 47-60 (2021-02-20)
Like bacterial and cytoplasmic ribosomes, mitoribosomes are large ribonucleoprotein complexes with molecular weights in the range of several million Daltons. Traditionally, studying the assembly of such high molecular weight complexes is done using ultracentrifugation through linear density gradients, which remains
Juliette Pulman et al.
Human molecular genetics, 28(9), 1445-1462 (2018-12-20)
Mitochondria contain a dedicated translation system, which is responsible for the intramitochondrial synthesis of 13 mitochondrial DNA (mtDNA)-encoded polypeptides essential for the biogenesis of oxidative phosphorylation (OXPHOS) complexes I and III-V. Mutations in nuclear genes encoding factors involved in mitochondrial
Sex-specific regulation of mitochondrial DNA levels: genome-wide linkage analysis to identify quantitative trait loci.
Lopez S
PLoS ONE, 7, e42711-e42711 (2012)
Reconstructing the evolution of the mitochondrial ribosomal proteome.
Smits P
Nucleic Acids Research, 35, 4686-4703 (2007)
MTO1 mediates tissue specificity of OXPHOS defects via tRNA modification and translation optimization, which can be bypassed by dietary intervention.
Tischner C
Human Molecular Genetics, 24, 2247-2266 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service