Skip to Content
Merck
All Photos(9)

Documents

HPA023900

Sigma-Aldrich

Anti-NFKB2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DNA-binding factor KBF2, Anti-H2TF1, Anti-Lymphocyte translocation chromosome 10, Anti-Lyt10, Anti-Nuclear factor NF-kappa-B p100 subunit, Anti-Oncogene Lyt-10

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ICNYEGPAKIEVDLVTHSDPPRAHAHSLVGKQCSELGICAVSVGPKDMTAQFNNLGVLHVTKKNMMGTMIQKLQRQRLRSRPQGLTEAEQRELEQEAKELKKVMDLSIVRLRFSAFLRASDGSFSLPLKPVISQP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKB2(4791)

General description

Nuclear factor-κ−B 2 (nfκb2) is a proto-oncogene mapped to human chromosome 10q24.1. The gene codes for NFκB2 subunit, also referred to as p100-p52, which is a member of NF-κB/Rel family of proteins. The encoded protein is characterized with a DNA-binding rel domain at its N-terminal end, a poly (G) hinge and an ankyrin domain at its C-terminal end. The native nfκb2 gene is localized to cytoplasm whereas the abnormal gene, which lacks IκB-like properties is expressed in nucleus.

Immunogen

Nuclear factor NF-kappa-B p100 subunit recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Nuclear factor -κ-B 2 (NFκB2) is the primary protein that takes part in the noncanonical NF-κB pathway and it also aids in various cellular functions such as peripheral lymphoid organ development, B cell development, and antibody production. NFκB-2 can act as either κB-mediated transcription activator or repressor depending on the relative concentration of its dimerization partner in the nucleus.
Aberrations in the C-terminal end of the NFκB2 gene contribute to the pathogenesis of various hematopoietic tumors, such as chronic lymphocytic leukemia, multiple myeloma, and cutaneous T-cell lymphoma gene. Mutations in NFκB2 lead to a heterogeneous disorder named common variable immunodeficiency (CVID).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86755

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rearranged NFKB-2 genes in lymphoid neoplasms code for constitutively active nuclear transactivators.
Chang cc
Molecular and Cellular Biology, 15(9), 5180-5187 (1995)
Germline mutations in NFKB2 implicate the noncanonical NF-?B pathway in the pathogenesis of common variable immunodeficiency.
Chen K
American Journal of Human Genetics, 93(5), 812-824 (2013)
M Heusch et al.
Oncogene, 18(46), 6201-6208 (1999-12-22)
nfkb2 encodes two members of the NF-kappa B/Rel family of proteins: p52 and p100. The p100 polypeptide has been proposed to serve as a precursor of p52, which corresponds to the N-terminal half of p100. While p52 functions as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service