Skip to Content
Merck
All Photos(6)

Documents

HPA022905

Sigma-Aldrich

Anti-SEPT4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Bradeion beta, Anti-Brain protein H5, Anti-CE5B3 beta, Anti-Cell division control-related protein 2, Anti-Cerebral protein 7, Anti-Peanut-like protein 2, Anti-Septin-4, Anti-hCDCREL-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEPTIN4(5414)

General description

SEPT4 (septin4) belongs to the septin family of GTPases and the gene is mapped to human chromosome 17q23. It is expressed in liver, brain, and testes.

Immunogen

Septin-4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SEPT4 (septin4) participates in cell apoptosis, cell cycle regulation and fibrogenesis. It is a Notch target gene. Changes in expression of SEPT4 might be associated with idiopathic asthenozoospermia. It is also responsible for TGF (transforming growth factor)-β-mediated apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75545

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Neubauer et al.
Cytoskeleton (Hoboken, N.J.), 76(1), 143-153 (2018-07-19)
Chronic kidney disease (CKD) is characterized by the loss of nephrons and worsening organ-fibrosis that leads to deterioration and ultimately the total breakdown of kidney function. Renal fibrosis has become a major public health problem worldwide and necessitates hemodialysis and
Katharina Neubauer et al.
Cytoskeleton (Hoboken, N.J.), 76(1), 143-153 (2018-07-19)
Chronic kidney disease (CKD) is characterized by the loss of nephrons and worsening organ-fibrosis that leads to deterioration and ultimately the total breakdown of kidney function. Renal fibrosis has become a major public health problem worldwide and necessitates hemodialysis and
Dandan Zhu et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 63(3), 163-169 (2014-12-21)
Apoptosis of activated hepatic stellate cells (HSCs) has been verified as a potential mechanism to aid in hepatic fibrosis remission. Earlier research suggests that Septin4_i1 may sensitize hepatocellular carcinoma cells to serum starvation-induced apoptosis. Here, we aimed to investigate the
Wenbin Liu
Molecular biology reports, 39(4), 4401-4409 (2011-09-23)
Notch receptor-mediated signaling is an evolutionarily conserved pathway that regulates diverse developmental processes and its dysregulation has been implicated in a variety of developmental disorders and cancers. Notch functions in these processes by activating expression of its target genes. Septin
Yu-Shan Li et al.
Zhonghua nan ke xue = National journal of andrology, 17(8), 699-702 (2011-09-09)
To investigate the role of the SEPT4 protein in the pathogenesis of idiopathic asthenozoospermia. Samples of ejaculated sperm from idiopathic asthenozoospermia patients and normozoospermic men were separated and purified by Percoll discontinuous density gradients, the distribution and expression of SEPT4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service