Skip to Content
Merck
All Photos(5)

Key Documents

HPA010687

Sigma-Aldrich

Anti-RHOT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MIRO-1 antibody produced in rabbit, Anti-Mitochondrial ρ-GTPase 1 antibody produced in rabbit, Anti-Rac-GTP-binding protein-like protein antibody produced in rabbit, Anti-Ras homolog gene family member T1 antibody produced in rabbit, Anti-hMiro-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

THIVDYSEAEQSDEQLHQEISQANVICIVYAVNNKHSIDKVTSRWIPLINERTDKDSRLPLILVGNKSDLVEYSSMETILPIMNQYTEIE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOT1(55288)

Related Categories

General description

RHOT1 is a protein found mainly in mitochondria. It has a pivotal role in mitochondrial homeostasis and apoptosis. It is also involved in membrane trafficking. Anti-RHOT1 antibody can be used in protein array. Rabbit anti-RHOT1 antibody reacts specifically with RHOT1 protein.

Immunogen

Mitochondrial ρ-GTPase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71907

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferdinand F Fatiga et al.
Journal of cellular biochemistry, 122(12), 1848-1862 (2021-08-26)
Mitochondria function as an integrated network that moves along the microtubules within cells and changes the morphology through membrane fusion and fission events. Mitofusin (MFN) mediates membrane tethering and subsequent fusion of the mitochondrial outer membrane. Understanding the regulatory mechanisms
Bing Zhou et al.
The Journal of cell biology, 214(1), 103-119 (2016-06-09)
Although neuronal regeneration is a highly energy-demanding process, axonal mitochondrial transport progressively declines with maturation. Mature neurons typically fail to regenerate after injury, thus raising a fundamental question as to whether mitochondrial transport is necessary to meet enhanced metabolic requirements
Asa Fransson et al.
The Journal of biological chemistry, 278(8), 6495-6502 (2002-12-17)
The human genomic sequencing effort has revealed the presence of a large number of Rho GTPases encoded by the human genome. Here we report the characterization of a new family of Rho GTPases with atypical features. These proteins, which were
Vinita Bharat et al.
Frontiers in cell and developmental biology, 9, 765408-765408 (2021-11-23)
Mutations in MAPT gene cause multiple neurological disorders, including frontal temporal lobar degeneration and parkinsonism. Increasing evidence indicates impaired mitochondrial homeostasis and mitophagy in patients and disease models of pathogenic MAPT. Here, using MAPT patients' fibroblasts as a model, we
Sa Fransson et al.
Biochemical and biophysical research communications, 344(2), 500-510 (2006-04-25)
We recently described the atypical Rho GTPases Miro-1 and Miro-2. These proteins have tandem GTP-binding domains separated by a linker region with putative calcium-binding motives. In addition, the Miro GTPases have a C-terminal transmembrane domain, which confers targeting to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service