Skip to Content
Merck
All Photos(4)

Documents

HPA002381

Sigma-Aldrich

Anti-DPYSL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CRMP-2, Anti-Collapsin response mediator protein 2, Anti-DRP-2, Anti-Dihydropyrimidinase-related protein 2, Anti-N2A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DFFQGTKAALAGGTTMIIDHVVPEPGTSLLAAFDQWREWADSKSCCDYSLHVDISEWHKGIQEEMEALVKDHGVNSFLVYMAFKDRFQLTDCQIYEVLSVIRDIGAIAQVHAENGDIIAEEQQRILDLGITGPEGHVLSR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPYSL2(1808)

General description

Dihydropyrimidinase-like 2 (DPYSL2) belongs to the CRMP/TOAD/Ulip/DRP family of cytosolic phosphoproteins associated with neuronal differentiation and axonal guidance.

Immunogen

Dihydropyrimidinase-related protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Dihydropyrimidinase-like 2 (DPYSL2) plays a vital role in neuronal development as well as polarity. It also helps in primary axon growth, axon induction in hippocampal neurons. It has a major role in the regulation of endocytic transport in non-neuronal cells. It interacts with endocytic transport regulatory protein MICAL-L1. DPYSL2 also negatively regulates dynein-based transport in neuronal cells by interacting with tubulin dimers and kinesin. It is essential for collapsin 1/semaphorin 3A signaling in intracellular response for axonal outgrowth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86191

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Inagaki et al.
Nature neuroscience, 4(8), 781-782 (2001-07-31)
In cultured hippocampal neurons, one axon and several dendrites differentiate from a common immature process. Here we found that CRMP-2/TOAD-64/Ulip2/DRP-2 (refs. 2-4) level was higher in growing axons of cultured hippocampal neurons, that overexpression of CRMP-2 in the cells led
Mina Noura et al.
Scientific reports, 10(1), 20245-20245 (2020-11-22)
Although the biological importance of Krüppel-like factor 4 (KLF4) transcription factor in the terminal differentiation of hematopoietic cells to the monocytes has been well established, the underlying mechanisms remain elusive. To clarify the molecular basis of KLF4-mediated monocytic differentiation, we
Juliati Rahajeng et al.
The Journal of biological chemistry, 285(42), 31918-31922 (2010-08-31)
Endocytosis is a conserved cellular process in which nutrients, lipids, and receptors are internalized and transported to early endosomes, where they are sorted and either channeled to degradative pathways or recycled to the plasma membrane. MICAL-L1 and EHD1 are important
Y Gu et al.
The Journal of biological chemistry, 275(24), 17917-17920 (2000-04-20)
Collapsin response mediator protein-2 (CRMP-2) is a member of the CRMP/TOAD/Ulip/DRP family of cytosolic phosphoproteins involved in neuronal differentiation and axonal guidance. CRMP-2 mediates the intracellular response to collapsin 1/semaphorin 3A, a repulsive extracellular guidance cue for axonal outgrowth. The
Alja Zottel et al.
Therapeutic advances in medical oncology, 12, 1758835920915302-1758835920915302 (2020-05-20)
Glioblastoma is a particularly common and very aggressive primary brain tumour. One of the main causes of therapy failure is the presence of glioblastoma stem cells that are resistant to chemotherapy and radiotherapy, and that have the potential to form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service