Skip to Content
Merck
All Photos(1)

Key Documents

GE29400456

HiTrap® Capto Adhere

Cytiva 29400456, pack of 1 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NB.13

packaging

pack of 1 × 1 mL

manufacturer/tradename

Cytiva 29400456

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

HiTrap Capto adhere is prepacked with Capto adhere, a multimodal BioProcess ion exchange resin designed to remove contaminats in post-protein A purification. Capto adhere is a strong anion exchanger with multimodal functionality. The multimodal functionality gives a different selectivity compared to traditional anion exchangers. Capto adhere is designed for post-protein A purification of monoclonal antibodies. Removal of leached protein A, aggregates, host cell proteins, nucleic acids, and viruses from monoclonal antibodies is performed in flowthrough mode at which the antibodies pass directly through the column while the contaminants are adsorbed. For optimal performance of Capto adhere, screening/optimization of loading conditions are required.

Features and Benefits

  • High capacity and productivity in flowthrough mode.
  • Savings in time and operating costs with a two-step chromatographic process.
  • Wide operational window of pH and conductivity.
  • Prepacked HiTrap columns for easy screening and convenient process development.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service