Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9893-32

Sigma-Aldrich

HiLoad® 26/600 Superdex® 30 pg

Cytiva 28-9893-32, column L × I.D. 60 cm × 26 mm, 34 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

bed support (polyamide)
glass column
glass fiber (polypropylene reinforced)

mol wt

<10000 Da (globular proteins)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9893-32

parameter

<13 mL sample volume
<2.6 mL/min flow rate
0.3 Mpa (42 psi)

bed volume

320 mL

column L × I.D.

60 cm × 26 mm

matrix

agarose-dextran composite
dextran-agarose particle platform
polymeric

matrix active group

dextran-agarose phase
dextran-agarose phase

avg. part. size

34 μm

cleaning

1-14

working range

3-12

separation technique

size exclusion (SEC)

General description

HiLoad 26/600 Superdex 30 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography separating peptides.

Suitability

All commonly used buffers, 1 M NaOH, 0.1 M HCl, 1 M acetic acid, 8 M urea, 6 M guanidine HCl, 1 % SDS, 70 % ethanol, 30 % iso-propanol, 30 % acetonitrile
for high-resolution preparative gel filtration chromatography separating peptides

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Legal Information

HiLoad is a registered trademark of Cytiva
Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Understanding pressure drop in chromatography systems is crucial for optimal functionality across different parts of the system.

Protocols

Superdex from Cytiva are SEC media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a superdex column.

Ensure chromatographic sample clarity to avoid column clogging, reduce washing needs, and prolong column life for purification.

Ensure chromatographic sample clarity to avoid column clogging, reduce washing needs, and prolong column life for purification.

Ensure chromatographic sample clarity to avoid column clogging, reduce washing needs, and prolong column life for purification.

See All

Related Content

Superdex® from Cytiva are Size Exclusion Chromatography media consisting of a composite base matrix of dextran and agarose. This page shows how to perform a separation with a Superdex® column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service