GE10600120
Amersham™ Protran® Supported Western blotting membrane sandwich, nitrocellulose
pore size 0.2 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600120
sheet W × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
General description
Amersham™ Protran® Supported Sandwich NC contains 10 Western blotting sandwiches, consisting of precut Amersham™ Protran® Supported nitrocellulose membranes preassembled with 2 x 3MM Chr filter papers. Designed for convenience and ease-of-use.
Features and Benefits
- Precut Amersham Protran 0.2 µm supported nitrocellulose membranes.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service