EP5820708000
Wide Neck Bottle for Eppendorf® S-4-104 Rotor
round, pack of 2, capacity 750 mL
Synonym(s):
Lid for Eppendorf® FA-45-48-11 Rotor
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Recommended Products
material
polypropylene bottle
round
feature
with cap
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5820708000 (Global)
parameter
3250 × g max. RCF
capacity
750 mL
Related Categories
Features and Benefits
- The bottle is made of polypropylene and supplied with a screw cap
- Compatible for rotors S-4-104, S-4x750, S-4x1000
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)