Skip to Content
Merck
All Photos(1)

Key Documents

EP0030602102

Eppendorf® Microplate 96/F-PP

clear wells, white border, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
colorless wells
polypropylene
size:
96 well
sterility:
sterile; autoclaved
sterile

material

colorless wells
polypropylene

sterility

sterile; autoclaved
sterile

packaging

pkg of 80 plates (5 bags x 16 plates )

manufacturer/tradename

Eppendorf® 0030602102

technique(s)

PCR: suitable

size

96 well

color

clear wells
white border

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service