EP0030128656
Eppendorf® Twin tec PCR plates, 96 wells
yellow plate, conical bottom colorless wells, skirted, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
polycarbonate
yellow plate
polycarbonate
yellow plate
size:
96 wells
feature:
skirt (Skirted)
Recommended Products
material
conical bottom colorless wells
polycarbonate
yellow plate
feature
skirt (Skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128656
technique(s)
PCR: suitable
size
96 wells
well maximum volume
40 μm
Looking for similar products? Visit Product Comparison Guide
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Features and Benefits
- Eppendorf® Twin tec PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and extremely robust polycarbonate frames for ultimate rigidity and torque-resistance.
- Optimized rim for barcodes and automated work processes
- Within SBS footprint recommendations (127.76×85.47 mm±0.25 mm).
- Stackable.
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block.
- Low profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes.
- 150 μL max. well volume (when used with cap strips).
- Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service