Skip to Content
Merck
All Photos(1)

Key Documents

E1782

Sigma-Aldrich

Anti-EDEM2 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ER degradation enhancer, mannosidase alpha-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 0.5-1.0 μg/mL using whole extracts of human HepG2 and rat NRK cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... EDEM2(55741)
mouse ... Edem2(108687)
rat ... Edem2(296304)

General description

Three EDEM homologs, EDEM1, EDEM2 and EDEM3 have been identified, which are transcriptionally upregulated upon ER stress by the activated IRE1/Xbp-1 branch. EDEM2 is localized to the ER, mainly as a soluble glycoprotein, interacts with calnexin and lacks mannosidase activity.

Application

Anti-EDEM2 antibody produced in rabbit is suitable for indirect immunofluorescence and immunoblotting at a working concentration of 0.5-1.0μg/mL using whole extracts of human HepG2 and rat NRK cells.

Biochem/physiol Actions

EDEM2 (ER degradation-enhancing α-mannosidase-like protein 2), a stress-regulated mannosidase-like protein, targets misfolded glycoproteins for degradation in an N-glycan dependent manner. Over-expression of EDEM2 accelerates ERAD (ER-associated degradation) by promoting the release of terminally misfolded glycoproteins from the calnexin cycle, without affecting the rate of degradation of non-glycosylated polypeptides or the maturation of model secretory proteins.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristian V A Munteanu et al.
Molecular & cellular proteomics : MCP, 20, 100125-100125 (2021-08-01)
Various pathologies result from disruptions to or stress of endoplasmic reticulum (ER) homeostasis, such as Parkinson's disease and most neurodegenerative illnesses, diabetes, pulmonary fibrosis, viral infections, and cancers. A critical process in maintaining ER homeostasis is the selection of misfolded
Silvia Olivari et al.
The Journal of biological chemistry, 280(4), 2424-2428 (2004-12-08)
Proteins expressed in the endoplasmic reticulum (ER) are subjected to a tight quality control. Persistent association with ER-resident molecular chaperones prevents exit of misfolded or incompletely assembled polypeptides from the ER and forward transport along the secretory line. ER-associated degradation
Steven W Mast et al.
Glycobiology, 15(4), 421-436 (2004-11-13)
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). Early in this pathway, a proposed lumenal ER lectin, EDEM, recognizes misfolded glycoproteins in the
Cristina Pintado et al.
Scientific reports, 7(1), 8100-8100 (2017-08-16)
Proteostasis alteration and neuroinflammation are typical features of normal aging. We have previously shown that neuroinflammation alters cellular proteostasis through immunoproteasome induction, leading to a transient decrease of proteasome activity. Here, we further investigated the role of acute lipopolysaccharide (LPS)-induced
Taiki Kuribara et al.
Chembiochem : a European journal of chemical biology, 18(11), 1027-1035 (2017-04-04)
Within the endoplasmic reticulum, immature glycoproteins are sorted into secretion and degradation pathways through the sequential trimming of mannose residues from Man9 GlcNAc2 to Man5 GlcNAc2 by the combined actions of assorted α-1,2-mannosidases. It has been speculated that specific glycoforms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service