CLS431444
Corning® cap, Erlenmeyer access, LUER, 1 L
Synonym(s):
Erlenmeyer flask cap
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
manufacturer/tradename
Corning 431444
availability
available only in South Korea
Related Categories
General description
- Aseptic transfer caps are chemically resistant and fit Corning® Erlenmeyer 1L flasks.
- Features a venting port that ends in a 50mm diameter vent filter with a 0.2 μm pore Polyvinylidene fluoride (PVDF) membrane and a barbed fitting.
- The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask.
- Heat sealable flexible tubing ends in a male Luer lock (MLL) closed with a female Luer sealing cap.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)