Skip to Content
Merck
All Photos(2)

Key Documents

CLS431131

Corning® cryogenic boxes, racks & trays

Reusable vial rack for 50 vials

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122806
NACRES:
NB.13
Pricing and availability is not currently available.

material

polypropylene

sterility

non-sterile

feature

Holds 50 x cryovials

packaging

pack of 2 ea

manufacturer/tradename

Corning 431131

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Reusable racks are designed for use with cryogenic vials.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service