Skip to Content
Merck
All Photos(1)

Key Documents

CLS3885

Corning® 96 Well Half-Area Microplate

flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear
round wells
white polystyrene
size:
96 wells
well D:
10.7 mm
well volume:
190 μL
well working volume:
25-125 μL
Pricing and availability is not currently available.

material

flat bottom clear
round wells
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

bag of 20
pkg of 25 inner package

manufacturer/tradename

Corning 3885

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

white

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate has half the growth area of a standard 96 well plate.
  • Flat bottoms with 190 μL total volume.
  • White walled plates enhance luminescent signals and have low background luminescence and fluorescence.
  • Bottoms are 60% thinner than conventional polystyrene plates.
  • Opaque walls to prevent well-to-well crosstalk.
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments.
  • Sterilized by gamma radiation and nonpyrogenic.
  • Hydrophilic surface and negatively charged, optimized for cell attachment and growth.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service