Skip to Content
Merck
All Photos(1)

Key Documents

CLS353046

Corning® Falcon® Cell Culture Plate

wells, 6, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile

Synonym(s):

Cell Culture Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

flat bottom clear wells

sterility

sterile

feature

lid
wells 6

packaging

case of 50 ea (Individually wrapped)

manufacturer/tradename

Corning 353046

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® 6 Well Clear Flat Bottom, Tissue Culture-Treated Multiwell Cell Culture Plate with Lid, Sterile, Individually Wrapped, 50/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service