Skip to Content
Merck
All Photos(1)

Key Documents

Nthy-ts1

90011610, human thyroid, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human thyroid
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified

Product Name

Nthy-ts1, 90011610

biological source

human thyroid

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human thyroid follicular mutant epithelial

Cell Line Description

Normal primary cultures of human thyroid follicular cells were transfected with a plasmid containing the SV40 early region large T encoding gene from mutant tsA58. Tight temperature dependence for growth is observed: Nthy-ts1 cells grow at 33°C, but not at the restrictive temperature of 40.5°C. This SV40 mutant allows the study of oncogene action free of any interfering effects of SV40 large T antigen.

Application

Transfection, mutation and virus studies: SV40

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 33°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service