BMSD113450
Benchmark Stainless Steel Bulk Beads
bead diam. 5 mm, acid washed
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
Recommended Products
material
stainless steel
packaging
pack of 1000 ea
manufacturer/tradename
Benchmark Scientific D1134-50
bead diam.
5 mm
General description
Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free
Choose from one of the most recent versions:
Certificates of Analysis (COA)