Skip to Content
Merck
All Photos(1)

Key Documents

AXYT300STKS

Pipette tips, for P2/P10

stacked racks, size 0.5-10 μL, clear, pkg of 10x480tips/cs, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
size:
0.5-10 μL
sterility:
sterile
packaging:
pkg of 10x480tips/cs
manufacturer/tradename:
Axygen Inc T-300-STK-S
Corning

sterility

sterile

manufacturer/tradename

Axygen Inc T-300-STK-S
Corning

packaging

pkg of 10x480tips/cs

size

0.5-10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service