Skip to Content
Merck
All Photos(1)

Key Documents

18723

Sigma-Aldrich

Mant-ATP triethylammonium salt solution

≥95.0% (HPLC)

Synonym(s):

2′/3′-O-(N-Methylanthraniloyl)adenosine 5′-triphosphate triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23N6O14P3 · xC6H15N × yH2O
CAS Number:
Molecular Weight:
640.33 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(OP(O)(OP(O)(OC[C@@H]1[C@@H]([R1])[C@@H]([R2])[C@H](N(C=N2)C3=C2C(N)=NC=N3)O1)=O)=O)(O)=O.CCN(CC)CC.O=C(C4=C(NC)C=CC=C4)O.O=C(C5=C(NC)C=CC=C5)O

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kateřina Skopalová et al.
International journal of molecular sciences, 22(2) (2021-01-10)
The active role of biomaterials in the regeneration of tissues and their ability to modulate the behavior of stem cells in terms of their differentiation is highly advantageous. Here, polypyrrole, as a representantive of electro-conducting materials, is found to modulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service