Skip to Content
Merck
All Photos(1)

Key Documents

Y0000530

Sultamicillin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N4O9S2
CAS Number:
Molecular Weight:
594.66
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sultamicillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C25H30N4O9S2/c1-24(2)17(29-20(32)16(21(29)39-24)27-19(31)15(26)12-8-6-5-7-9-12)22(33)37-11-38-23(34)18-25(3,4)40(35,36)14-10-13(30)28(14)18/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31)/t14?,15-,16?,17-,18-,21?/m0/s1

InChI key

OPYGFNJSCUDTBT-CUUJHUNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficacy and safety of Sultamicillin (Ampicillin/Sulbactan) and Amoxicillin/Clavulanic Acid in the treatment of upper respiratory tract infections in adults: an open-label, multicentric, randomized trial.
Ferreira J B, et al.
Revista Brasileira de Otorrinolaringologia, 72(1), 104-111 (2006)
HPLC for in-process control in the production of sultamicillin.
Laviana L, et al.
Journal of Pharmaceutical and Biomedical Analysis, 31(2), 321-328 (2003)
Efficacy and safety of sultamicillin (750 mg bid) compared with amoxycillin/clavulanate (625 mg tid) in patients with umcomplicated urinary tract infections.
Schutz W.
International Journal of Antimicrobial Agents, 6, S55-S59 (1996)
Ralf Smeets et al.
The International journal of oral & maxillofacial implants, 32(6), 1338–1345-1338–1345 (2017-10-14)
To test the antimicrobial properties, surface topography, reaction of surrounding tissue (biocompatibility), and osseointegration of ultrathin implant surfaces containing polysiloxane and nanoscaled silver particles. Implants with polysiloxane coating and nanoscaled silver particles (Ag/SiO Roughness parameters did not differ between all
Mahir Seyrek et al.
Journal of endourology, 26(11), 1431-1436 (2012-05-23)
To compare sulbactam-ampicillin and cefuroxime antibiotics for prophylaxis of percutaneous nephrolithotomy (PCNL) and to find out the optimal regimen for antibiotic maintenance to prevent systemic inflammatory response syndrome (SIRS). Between February 2010 and March 2011, a total of 198 patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service