Skip to Content
Merck
All Photos(1)

Key Documents

W314820

Sigma-Aldrich

Ethyl 2-trans-4-cis-decadienoate

≥95%, stabilized, FG

Synonym(s):

Ethyl (2E,4Z)-deca-2,4-dienoate, Ethyl (E,Z)-2,4-decadienoate, Pear ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH=CHCO2C2H5
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3148
Beilstein:
1724176
EC Number:
Council of Europe no.:
10574
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.260
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥95%

contains

α-tocopherol synthetic as stabilizer

refractive index

n20/D 1.486 (lit.)

bp

70-72 °C/0.05 mmHg (lit.)

density

0.905 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; tropical; waxy; fruity; pear; pear; sweet

SMILES string

[H]\C(CCCCC)=C([H])\C([H])=C\C(=O)OCC

InChI

1S/C12H20O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h8-11H,3-7H2,1-2H3/b9-8-,11-10+

InChI key

OPCRGEVPIBLWAY-QNRZBPGKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service