Skip to Content
Merck
All Photos(1)

Key Documents

SMB01322

Sigma-Aldrich

Maltotetraose

94% (HPLC)

Synonym(s):

α-1,4-Tetraglucose, Amylotetraose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
Beilstein:
78870
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

Assay

94% (HPLC)

form

solid

color

white

shipped in

ambient

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]4CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-5-9(29)10(30)15(35)22(40-5)44-19-7(3-27)42-24(17(37)12(19)32)45-20-8(4-28)41-23(16(36)13(20)33)43-18-6(2-26)39-21(38)14(34)11(18)31/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10+,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21+,22-,23-,24-/m1/s1

InChI key

LUEWUZLMQUOBSB-ZLBHSGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltotetraose also known as Amylotetraose, is a tetrasaccharide that is composed of glucose molecules linked by α-1,4 glycosidic bonds and has been commonly found in B. stearothermophilus. It increases the α-amylase synthesis rate in B. stearothermophilus and inhibits TNF-α-induced expression of intercellular adhesion molecule-1 (ICAM-1). Maltotetraose can be used as a substrate for the enzyme-coupled determination of amylase activity in biological fluids that finds application in metabolomics and biochemical research.

Application

Maltotetraose producing amylases are highly desirable for application in bread making and other food industries. It has been used in a study to characterize an exo-acting intracellular a-amylase from the hyperthermophilic bacterium Thermotoga neapolitana.

Features and Benefits

  • High-purity compound suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung-Min Park et al.
Applied microbiology and biotechnology, 86(2), 555-566 (2009-10-17)
We cloned and expressed the gene for an intracellular alpha-amylase, designated AmyB, from the hyperthermophilic bacterium Thermotoga neapolitana in Escherichia coli. The putative intracellular amylolytic enzyme contained four regions that are highly conserved among glycoside hydrolase family (GH) 13 alpha-amylases.
Sumit Kumar et al.
Bioresource technology, 116, 247-251 (2011-12-27)
Maltooligosaccharides especially maltotriose and maltotetraose producing amylases are highly desirable for application in bread making and other food industries. A maltotriose and maltotetraose producing amylase from moderately halophilic Marinobacter sp. EMB8 is described. Under optimized culture conditions, 48.0 IU/mL amylase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service