Skip to Content
Merck
All Photos(1)

Key Documents

PHR1661

Supelco

Isopentane Solution

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Methylbutane, Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

form

liquid

CofA

current certificate can be downloaded

autoignition temp.

788 °F

expl. lim.

8.3 %

packaging

ampule of 2 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.354 (lit.)

bp

30 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

-10 to -25°C

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. ′These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

This a solution of Isopentane in n-Butanol (~13%w/w)

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3677 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aneesh Donde et al.
Autophagy, 16(4), 672-682 (2019-06-27)
A shared neuropathological hallmark in amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD) is nuclear clearance and cytoplasmic aggregation of TARDBP/TDP-43 (TAR DNA binding protein). We previously showed that the ability of TARDBP to repress nonconserved cryptic exons was impaired
Eva Pigna et al.
EBioMedicine, 40, 717-732 (2019-02-05)
Histone deacetylase 4 (HDAC4) has been proposed as a target for Amyotrophic Lateral Sclerosis (ALS) because it mediates nerve-skeletal muscle interaction and since its expression in skeletal muscle correlates with the severity of the disease. However, our recent studies on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service