Skip to Content
Merck
All Photos(1)

Key Documents

NIST1244

Inconel® 600

NIST® SRM® 1244

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

technique(s)

XRF: suitable
optical emission spectroscopy (OES): suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1244 contains a wrought disk with a diameter of 35 mm and a thickness of 19 mm.


SRM 1244_cert

SRM 1244_SDS

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction values, expanded uncertainty, coverage factor for total elements in Inconel® 600.
  • The reference and information values of elements are reported as mass fraction.
  • The certified values are metrologically traceable to the SI derived unit of mass fraction, expressed as percent of milligrams per kilogram.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Cobalt (Co), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Vanadium (V), Boron (B), Magnesium (Mg), Lead (Pb), Thallium (Tl)

Reference/Informational value is provided for the following analytes.
Carbon (C), Niobium (Nb), Phosphorus (P), Sulfur (S), Silicon (Si), Titanium (Ti), Arsenic (As), Tin (Sn), Zirconium (Zr), Silver (Ag), Bismuth (Bi), Calcium (Ca), Gallium (Ga), Nitrogen (N), Oxygen (O), Antimony (Sb), Selenium (Se), Tantalum (Ta), Tellurium (Te), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

Inconel is a registered trademark of Special Metals Corporation
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service