Skip to Content
Merck
All Photos(1)

Documents

N1400000

Nystatin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fungicidin, Mycostatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nystatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[C@@H]([C@H](C)[C@H](C)O1)[C@@H](C)/C=C/C=C/CC/C=C/C=C/C=C/C=C/[C@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@H](N)[C@@H]2O)C[C@@]3([H])[C@H](C(O)=O)[C@@H](O)C[C@](O)(O3)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)C[C@@H](O)CC1=O

InChI

1S/C47H75NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-36(53)35(52)20-19-31(49)21-32(50)22-33(51)23-39(55)62-29(3)28(2)42(27)56/h5-6,8,10-18,27-38,40-44,46,49-54,56-58,61H,7,9,19-26,48H2,1-4H3,(H,59,60)/b6-5+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28+,29-,30+,31+,32+,33+,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

VQOXZBDYSJBXMA-QEKUPDCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nystatin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mode of Action: Nystatin acts by binding to ergosterols in fungal cell membranes, increasing permeability of the membrane, and creating nystatin/ergosterol based ion channels. It also functions as a lipid raft-inhibiting reagent and as membrane associated cholesterol.

Antimicrobial spectrum: Nystatin acts against fungi, yeasts and molds.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Giuliano et al.
Annali di igiene : medicina preventiva e di comunita, 25(1), 57-64 (2013-02-26)
The present study represents a preliminary evaluation of the impact of topical nystatin prophylaxis administration and adequate CVC management on reducing the chance of developing candidemia in neurosurgical intensive care unit (NICU) patients. We conducted a case-control study at the
Santhanam Shanmughapriya et al.
Biofouling, 28(9), 929-935 (2012-09-12)
The presence of intrauterine contraceptive devices (IUDs) gives a solid surface for attachment and an ideal niche for biofilm to form and flourish. Pelvic actinomycosis is often associated with the use of IUDs. Treatment of IUD-associated pelvic actinomycosis requires the
Ravikumar B Shinde et al.
Annals of clinical microbiology and antimicrobials, 11, 27-27 (2012-10-06)
Biofilms formed by Candida albicans are resistant towards most of the available antifungal drugs. Therefore, infections associated with Candida biofilms are considered as a threat to immunocompromised patients. Combinatorial drug therapy may be a good strategy to combat C. albicans
Ginekologia polska, 83(12), 956-959 (2013-03-16)
The group of experts representing the Polish Gynecologic Society has issued this statement based on the review of available literature on the potential benefits of the use of Macmiror Complex 500 in obstetrical and gynecologic practice. Mixed Vaginitis (MV) eg.
E N Gromosova et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 74(5), 26-31 (2012-11-06)
Inactivation of the key genes, which are responsible for the enzymes of polyphosphates degradation, exopolyphosphatases ppx1 and ppn1, caused both an increase of polyphosphates content in Saccharomyces cerevisiae cells and an increase in chain length of acid-soluble and alkali-soluble fractions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service