Skip to Content
Merck
All Photos(1)

Key Documents

CN34892

Sigma-Aldrich

Acetonitrile

Preparateur, ≥99.9% (GC), Customized plastic drum

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

GC/GC: suitable

impurities

≤0.005% free acid (as CH3COOH)
≤0.02% water (Karl Fischer)
≤1 mg/L non-volatile matter

color

colorless

transmittance

200 nm, >50%
220 nm, >90%

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (MeCN), an aliphatic nitrile, is a colorless liquid with a characteristic odor. MeCN has low viscosity, high elution strength and is highly miscible in water. It is widely used as a solvent and intermediate in organic syntheses and is transparent to UV-visible light, making it highly applicable in spectrophotometric and fluorimetric techniques. It also plays a major role as an extractant medium in liquid-liquid extraction, solid-phase extraction or microextraction.

Application

Acetonitrile may be used as solvent in the following processes:
  • Organic synthesis
  • Liquid-liquid extraction, solid-phase extraction or microextraction.
  • Spectrophotometric and fluorimetric techniques.
  • Electrolytes in lithium-ion batteries
  • Determination of pKa values of organic superbases using the isodensity polarization continuum model (IPCM).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Oxidative Arylalkylation of Activated Alkenes: Dual C-H Bond Cleavage of an Arene and Acetonitrile.
Wu T, et al.
Angewandte Chemie (International Edition in English), 50(52), 12578-12581 (2011)
Liquid phase hydrogenation of acetonitrile to ethylamine over the Co-B amorphous alloy catalyst.
Li H, et al.
J. Catal., 214(1), 15-25 (2003)
Hydrogen peroxide oxygenation of alkanes including methane and ethane catalyzed by iron complexes in acetonitrile.
Shul'pin GB, et al.
Advanced Synthesis & Catalysis, 346(2-3), 317-332 (2004)
Basicity of some organic superbases in acetonitrile.
Kovacevic B and Maksic ZB.
Organic Letters, 3(10), 1523-1526 (2001)
Dynamics of ultrafast intramolecular charge transfer with 4-(dimethylamino) benzonitrile in acetonitrile.
Druzhinin SI, et al.
The Journal of Physical Chemistry A, 110(9), 2955-2969 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service