Skip to Content
Merck
All Photos(2)

Key Documents

45628

Supelco

Pirimiphos-ethyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N3O3PS
CAS Number:
Molecular Weight:
333.39
Beilstein:
7141373
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1cc(C)nc(n1)N(CC)CC

InChI

1S/C13H24N3O3PS/c1-6-16(7-2)13-14-11(5)10-12(15-13)19-20(21,17-8-3)18-9-4/h10H,6-9H2,1-5H3

InChI key

TZBPRYIIJAJUOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pirimiphos-ethyl belongs to the class of organophosphorus pesticides, with contact and respiratory action and is commonly employed for the protection of crops including vegetables, bananas, turf and mushrooms. Its mode of action involves the inhibition of acetylcholinesterase activity.

Application

Pirimiphos-ethyl may be used as an analytical reference standard for the quantification of the analyte in honey bees and biological samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Determination of organophosphorus pesticides in honeybees after solid-phase microextraction
Fernandez M, et al.
Journal of Chromatography A, 922(1-2), 257-265 (2001)
[Severe suicidal poisoning with the preparation Primicid 20 during pregnancy].
I Domańska et al.
Przeglad lekarski, 40(6), 509-510 (1983-01-01)
Multiresidue determination method for organophosphorus pesticides in serum and whole blood by gas chromatography-mass-selective detection.
Lacassie E, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 759(1), 109-116 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service