41612
PCB No 209 solution
10±0.5 μg/mL in isooctane, analytical standard
Synonym(s):
Decachlorobiphenyl solution
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (GC)
shelf life
limited shelf life, expiry date on the label
concentration
10±0.5 μg/mL in isooctane
refractive index
n/D 1.3900-1.3940
format
single component solution
storage temp.
2-8°C
SMILES string
Clc1c(Cl)c(Cl)c(c(Cl)c1Cl)-c2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl
InChI
1S/C12Cl10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16
InChI key
ONXPZLFXDMAPRO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service